siRNA Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206367.1 | NP_001193296.1 | 3 | 336 | ||
| NM_001206368.1 | NP_001193297.1 | 2 | 252 | |||
| NM_001206369.1 | NP_001193298.1 | 2 | 297 | |||
| NM_146120.4 | NP_666232.2 | 2 | 359 | |||
| GenBank mRNA | AK076156.1 | - | 2 | 299 | ||
| AK089934.1 | - | 3 | 291 | |||
| AK135632.1 | - | 1 | 72 | |||
| AK143664.1 | - | 2 | 326 | |||
| AK146969.1 | - | 3 | 253 | |||
| AK148957.1 | - | 3 | 316 | |||
| AK150359.1 | - | 3 | 259 | |||
| AK151684.1 | - | 3 | 278 | |||
| AK153029.1 | - | 3 | 256 | |||
| AK166831.1 | - | 2 | 272 | |||
| AK166932.1 | - | 3 | 250 | |||
| AK167918.1 | - | 3 | 266 | |||
| AK168461.1 | - | 3 | 269 | |||
| AK168532.1 | - | 3 | 270 | |||
| AK168707.1 | - | 2 | 205 | |||
| AK168725.1 | - | 3 | 253 | |||
| AK168829.1 | - | 2 | 304 | |||
| AK169041.1 | - | 3 | 256 | |||
| AK169670.1 | - | 3 | 281 | |||
| AK183123.1 | - | 3 | 301 | |||
| BC023143.1 | - | 2 | 297 | 3711602 | ||
| BC060377.1 | - | 3 | 284 | 4983664 | ||
| BY144206.1 | - | 2 | 297 | |||

Gsn
Genomic Map