siRNA Details
| Gene Symbol: Ikzf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011771.1 | NP_035901.1 | 8 | 1655 | ||
| GenBank mRNA | AF001293.1 | - | 7 | 1374 | ||
| AK037261.1 | - | 1 | 64 | |||
| AK038043.1 | - | 2 | 2118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikzf3| Gene Symbol: Ikzf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011771.1 | NP_035901.1 | 8 | 1655 | ||
| GenBank mRNA | AF001293.1 | - | 7 | 1374 | ||
| AK037261.1 | - | 1 | 64 | |||
| AK038043.1 | - | 2 | 2118 | |||