siRNA Details
| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172662.3 | NP_766250.2 | 3 | 280 | ||
| GenBank mRNA | AK028468.1 | - | 3 | 203 | ||
| AK038057.1 | - | 2 | 151 | |||
| AK054272.1 | - | 3 | 281 | |||
| AK163943.1 | - | 3 | 238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtdc1| Gene Symbol: Gtdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172662.3 | NP_766250.2 | 3 | 280 | ||
| GenBank mRNA | AK028468.1 | - | 3 | 203 | ||
| AK038057.1 | - | 2 | 151 | |||
| AK054272.1 | - | 3 | 281 | |||
| AK163943.1 | - | 3 | 238 | |||