siRNA Details
| Gene Symbol: Slc30a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290993.1 | NP_001277922.1 | 2 | 773 | ||
| NM_011774.3 | NP_035904.2 | 3 | 920 | |||
| GenBank mRNA | AF003747.1 | - | 3 | 721 | ||
| AK134245.1 | - | 3 | 898 | |||
| AK166106.1 | - | 2 | 775 | |||
| BC117997.1 | - | 3 | 778 | 40095367 | ||
| CV556160.1 | - | 1 | 84 | |||

Slc30a4
Genomic Map