siRNA Details
Gene Symbol: 4933409G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177651.3 | NP_808319.1 | 13, 14 | 880 | ||
GenBank mRNA | AK077128.1 | - | 13, 14 | 881 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933409G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177651.3 | NP_808319.1 | 13, 14 | 880 | ||
GenBank mRNA | AK077128.1 | - | 13, 14 | 881 |