siRNA Details
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172665.5 | NP_766253.2 | 5 | 772 | ||
| GenBank mRNA | AK038454.1 | - | 4 | 2287 | ||
| AK046805.1 | - | 5 | 773 | |||
| AK085215.1 | - | 5 | 671 | |||
| AK153649.1 | - | 5 | 725 | |||
| AK163553.1 | - | 5 | 702 | |||
| BC025480.1 | - | 4 | 513 | |||
| BC027196.1 | - | 5 | 695 | 4458562 | ||

Pdk1
Genomic Map