siRNA Details
| Gene Symbol: Sestd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175465.6 | NP_780674.1 | 8 | 651 | ||
| GenBank mRNA | AK047939.1 | - | 8 | 649 | ||
| BC013245.1 | - | 9 | 732 | |||
| BC044853.1 | - | 8 | 651 | 3978465 | ||
| BG082832.2 | - | 5 | 394 | |||
| CO425092.1 | - | 2 | 84 | |||

Sestd1
Genomic Map