siRNA Details
| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145526.2 | NP_663501.2 | 12 | 1687 | ||
| GenBank mRNA | AK140283.1 | - | 12 | 2955 | ||
| AK141847.1 | - | 12 | 1352 | |||
| BC023089.1 | - | 12 | 1340 | 3984739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2rx3| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145526.2 | NP_663501.2 | 12 | 1687 | ||
| GenBank mRNA | AK140283.1 | - | 12 | 2955 | ||
| AK141847.1 | - | 12 | 1352 | |||
| BC023089.1 | - | 12 | 1340 | 3984739 | ||