siRNA Details
| Gene Symbol: 1110051M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175123.4 | NP_780332.1 | 1, 2 | 84 | ||
| GenBank mRNA | AK045977.1 | - | 1, 2 | 85 | ||
| AK164320.1 | - | 1, 2 | 82 | |||
| BC032255.1 | - | 1, 2 | 104 | |||
| BC085239.1 | - | 1, 2 | 83 | 6440075 | ||
| BC089006.1 | - | 1, 2 | 64 | 6843427 | ||

1110051M20Rik
Genomic Map