siRNA Details
| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172668.3 | NP_766256.3 | 20 | 2854 | ||
| GenBank mRNA | AF247637.1 | - | 20 | 2684 | ||
| AK129224.1 | - | 6 | 645 | |||
| BC132240.1 | - | 20 | 2777 | 40130517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp4| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172668.3 | NP_766256.3 | 20 | 2854 | ||
| GenBank mRNA | AF247637.1 | - | 20 | 2684 | ||
| AK129224.1 | - | 6 | 645 | |||
| BC132240.1 | - | 20 | 2777 | 40130517 | ||