siRNA Details
| Gene Symbol: Zfp770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175466.4 | NP_780675.1 | 2 | 462 | ||
| GenBank mRNA | AK032578.1 | - | 2 | 463 | ||
| AK043843.1 | - | 2 | 500 | |||
| AK077547.1 | - | 2 | 463 | |||
| AK164226.1 | - | 2 | 445 | |||
| BC137653.1 | - | 1 | 375 | 8860672 | ||
| BC137654.1 | - | 1 | 375 | 8860673 | ||

Zfp770
Genomic Map