siRNA Details
| Gene Symbol: Bahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045523.1 | NP_001038988.1 | 2 | 175 | ||
| GenBank mRNA | AK129243.1 | - | 2 | 333 | ||
| AK152597.1 | - | 2 | 113 | |||
| BC100358.1 | - | 2 | 337 | 30848279 | ||
| BC141046.1 | - | 1 | 38 | 9055658 | ||
| BC145233.1 | - | 1 | 38 | 9053785 | ||
| BY261229.1 | - | 2 | 175 | |||

Bahd1
Genomic Map