siRNA Details
Gene Symbol: Frmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172673.3 | NP_766261.2 | 8, 9 | 949 | ||
GenBank mRNA | AK030883.1 | - | 8, 9 | 705 | ||
AK044270.1 | - | 8, 9 | 700 | |||
AK054484.1 | - | 8, 9 | 906 | |||
AK146383.1 | - | 7, 8 | 829 | |||
BC038623.1 | - | 8, 9 | 686 | |||
BC057549.1 | - | 5, 6 | 478 | 6416914 | ||
BC062889.1 | - | 8, 9 | 906 | 5698263 |
