siRNA Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029432.1 | NP_083708.1 | 25, 26 | 2310 | ||
| GenBank mRNA | AK133294.1 | - | 25, 26 | 2452 | ||
| AK143611.1 | - | 25, 26 | 2335 | |||
| AK186845.1 | - | 3, 4 | 256 | |||
| BC052447.1 | - | 25, 26 | 2310 | 6408647 | ||

4930402H24Rik
Genomic Map