siRNA Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289738.1 | NP_001276667.1 | 2, 3 | 435 | ||
| NM_177656.4 | NP_808324.1 | 2, 3 | 435 | |||
| GenBank mRNA | AK078482.1 | - | 2, 3 | 435 | ||
| BC137826.1 | - | 1, 2 | 312 | 8860845 | ||
| BC144943.1 | - | 1, 2 | 312 | |||
| BC144944.1 | - | 1, 2 | 312 | 9053492 | ||
| BC144945.1 | - | 1, 2 | 308 | |||

6820408C15Rik
Genomic Map