siRNA Details
| Gene Symbol: Pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004721.1 | NP_001004721.1 | 2 | 188 | ||
| GenBank mRNA | AK140955.1 | - | 2 | 184 | ||
| AK143517.1 | - | 2 | 188 | |||
| AK162913.1 | - | 163 | ||||
| AK171988.1 | - | 2 | 167 | |||
| BB839941.1 | - | 2 | 188 | |||
| BC120680.1 | - | 2 | 177 | 40129603 | ||
| BC120682.1 | - | 2 | 177 | 40129605 | ||
| CK334159.1 | - | 2 | 141 | |||

Pigu
Genomic Map