siRNA Details
| Gene Symbol: Phf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172674.2 | NP_766262.2 | 9 | 1382 | ||
| GenBank mRNA | AK033017.1 | - | 8 | 1181 | ||
| AK038573.1 | - | 9 | 1289 | |||
| AK045309.1 | - | 9 | 1301 | |||
| AK203634.1 | - | 2 | 183 | |||
| BC060121.1 | - | 9 | 1262 | 6413021 | ||

Phf20
Genomic Map