siRNA Details
| Gene Symbol: Phf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172674.2 | NP_766262.2 | 15 | 2311 | ||
| GenBank mRNA | AK033017.1 | - | 14 | 2110 | ||
| AK171777.1 | - | 1 | 166 | |||
| AK190572.1 | - | 2 | 59 | |||
| BC011337.1 | - | 3 | 270 | |||
| BC060121.1 | - | 14 | 2230 | 6413021 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107795, XM_006235404, XM_008762342.

Phf20
Genomic Map