siRNA Details
| Gene Symbol: Ralgapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291137.1 | NP_001278066.1 | 24 | 3721 | ||
| NM_001291138.1 | NP_001278067.1 | 23 | 3685 | |||
| GenBank mRNA | AK122470.1 | - | 19 | 2435 | ||
| BC138377.1 | - | 24 | 3623 | 8861397 | ||
| BC145260.1 | - | 23 | 3587 | |||
| CB519158.1 | - | 2 | 234 | |||

Ralgapb
Genomic Map