siRNA Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146129.3 | NP_666241.1 | 9 | 1162 | ||
| GenBank mRNA | AK089358.1 | - | 9 | 1163 | ||
| AK150740.1 | - | 8 | 1053 | |||
| BC031431.1 | - | 9 | 1131 | 4241283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcif1| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146129.3 | NP_666241.1 | 9 | 1162 | ||
| GenBank mRNA | AK089358.1 | - | 9 | 1163 | ||
| AK150740.1 | - | 8 | 1053 | |||
| BC031431.1 | - | 9 | 1131 | 4241283 | ||