siRNA Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146129.3 | NP_666241.1 | 14 | 1900 | ||
| GenBank mRNA | AK089358.1 | - | 14 | 1901 | ||
| BC031431.1 | - | 14 | 1869 | 4241283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcif1| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146129.3 | NP_666241.1 | 14 | 1900 | ||
| GenBank mRNA | AK089358.1 | - | 14 | 1901 | ||
| BC031431.1 | - | 14 | 1869 | 4241283 | ||