siRNA Details
| Gene Symbol: Cbln4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175631.3 | NP_783439.1 | 2, 3 | 1570 | ||
| GenBank mRNA | AK032406.1 | - | 2, 3 | 1074 | ||
| AK032621.1 | - | 2, 3 | 1573 | |||
| AY134663.1 | - | 2, 3 | 402 | |||
| BC094540.1 | - | 2, 3 | 1473 | 5708067 | ||
| BC132025.1 | - | 2, 3 | 668 | 40130302 | ||
| BC132027.1 | - | 2, 3 | 668 | 40130304 | ||

Cbln4
Genomic Map