siRNA Details
| Gene Symbol: Npepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213733.2 | NP_998898.1 | 8 | 1052 | ||
| GenBank mRNA | AK150313.1 | - | 8 | 1053 | ||
| AK156136.1 | - | 8 | 1053 | |||
| AK170509.1 | - | 8 | 1053 | |||
| BC023239.1 | - | 7 | 730 | |||
| BC055448.1 | - | 7 | 727 | |||
| BC069933.1 | - | 8 | 1047 | 4008091 | ||
| BC080707.1 | - | 8 | 925 | |||

Npepl1
Genomic Map