siRNA Details
| Gene Symbol: Taf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081092.1 | NP_001074561.1 | 12 | 2755 | ||
| GenBank mRNA | AK136225.1 | - | 5 | 599 | ||
| AY038601.1 | - | 12 | 1611 | |||
| BC026493.1 | - | 3 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf4a| Gene Symbol: Taf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081092.1 | NP_001074561.1 | 12 | 2755 | ||
| GenBank mRNA | AK136225.1 | - | 5 | 599 | ||
| AY038601.1 | - | 12 | 1611 | |||
| BC026493.1 | - | 3 | 341 | |||