siRNA Details
Gene Symbol: Ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173761.3 | NP_776122.1 | 3, 4 | 347 | ||
GenBank mRNA | AK033241.1 | - | 4, 5 | 437 | ||
AK046768.1 | - | 3, 4 | 361 | |||
AK075846.1 | - | 3, 4 | 326 | |||
AK172582.1 | - | 3, 4 | 321 | |||
BC061479.1 | - | 3, 4 | 318 | 6334535 | ||
BC065050.1 | - | 3, 4 | 347 | 6418891 |
