siRNA Details
| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145919.1 | NP_001139391.1 | 4 | 829 | ||
| NM_172677.3 | NP_766265.3 | 5 | 862 | |||
| GenBank mRNA | AK039176.1 | - | 5 | 925 | ||
| AK053736.1 | - | 5 | 897 | |||
| AK137245.1 | - | 5 | 809 | |||
| BC042627.1 | - | 1 | 379 | |||
| BC052631.1 | - | 1 | 179 | |||
| BC057158.1 | - | 5 | 889 | 5348717 | ||
| BC067040.1 | - | 4 | 800 | 6853196 | ||
| BC067042.1 | - | 5 | 795 | 6856248 | ||

Ythdf3
Genomic Map