siRNA Details
| Gene Symbol: Acad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172678.3 | NP_766266.3 | 11 | 1236 | ||
| GenBank mRNA | AK028571.1 | - | 10 | 1950 | ||
| AK031820.1 | - | 11 | 1168 | |||
| AK075984.1 | - | 11 | 1155 | |||
| AK145423.1 | - | 11 | 1144 | |||
| AK205472.1 | - | 1 | 54 | |||
| BC004737.1 | - | 5 | 482 | |||
| BC031137.1 | - | 11 | 1172 | 4971848 | ||
| BC032213.1 | - | 11 | 1177 | 5323723 | ||
| BC033277.1 | - | 11 | 1157 | 4241041 | ||

Acad9
Genomic Map