siRNA Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291044.1 | NP_001277973.1 | 1 | 57 | ||
| NM_153561.3 | NP_705789.1 | 1 | 57 | |||
| GenBank mRNA | AF453428.1 | - | 1 | 43 | ||
| AK145299.1 | - | 1 | 49 | |||
| BC027267.1 | - | 1 | 51 | 3500531 | ||

Nudt6
Genomic Map