siRNA Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291044.1 | NP_001277973.1 | 3 | 491 | ||
| NM_153561.3 | NP_705789.1 | 3 | 491 | |||
| GenBank mRNA | AF453427.1 | - | 3 | 600 | ||
| AF453428.1 | - | 3 | 477 | |||
| AK145299.1 | - | 3 | 483 | |||
| BC027267.1 | - | 3 | 485 | 3500531 | ||
| BC146000.1 | - | 3 | 485 | 40131303 | ||

Nudt6
Genomic Map