siRNA Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177662.2 | NP_808330.1 | 1 | 61 | ||
| GenBank mRNA | AK020735.1 | - | 1 | 55 | ||
| AK034490.1 | - | 1 | 61 | |||
| AK040071.1 | - | 1 | 35 | |||
| AK049470.1 | - | 1 | 34 | |||
| AK143683.1 | - | 1 | 77 | |||
| AK165930.1 | - | 1 | 23 | |||
| AK166103.1 | - | 1 | 30 | |||
| BC098219.1 | - | 1 | 35 | 1066724 | ||

Ctso
Genomic Map