siRNA Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177662.2 | NP_808330.1 | 4 | 477 | ||
| GenBank mRNA | AK034490.1 | - | 4 | 477 | ||
| AK049470.1 | - | 4 | 450 | |||
| AK143683.1 | - | 4 | 493 | |||
| AK165930.1 | - | 4 | 439 | |||
| AK166103.1 | - | 4 | 446 | |||
| BC044664.1 | - | 4 | 369 | |||
| BC098219.1 | - | 4 | 451 | 1066724 | ||

Ctso
Genomic Map