siRNA Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172682.3 | NP_766270.2 | 9 | 1743 | ||
| GenBank mRNA | AK036881.1 | - | 6 | 729 | ||
| AK076386.1 | - | 1 | 595 | |||
| BC026906.1 | - | 5 | 552 | |||
| BC043298.1 | - | 7 | 787 | |||
| BC094511.1 | - | 7 | 954 | |||

Fam160a1
Genomic Map