siRNA Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172682.3 | NP_766270.2 | 12 | 3470 | ||
| GenBank mRNA | AK036881.1 | - | 9 | 2456 | ||
| AK076386.1 | - | 4 | 2325 | |||
| BC026906.1 | - | 8 | 2279 | |||
| BC043298.1 | - | 10 | 2514 | |||
| BC094511.1 | - | 10 | 2681 | |||

Fam160a1
Genomic Map