siRNA Details
| Gene Symbol: Smg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178246.3 | NP_839977.2 | 7 | 829 | ||
| GenBank mRNA | AK129286.1 | - | 7 | 808 | ||
| AK143911.1 | - | 7 | 831 | |||
| AK155787.1 | - | 7 | 818 | |||
| BC024683.1 | - | 7 | 781 | 3992000 | ||
| BC100408.1 | - | 7 | 778 | 30734268 | ||

Smg5
Genomic Map