siRNA Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144898.2 | NP_659147.2 | 13 | 1463 | ||
| GenBank mRNA | AK134001.1 | - | 1066 | |||
| AK139708.1 | - | 1442 | ||||
| AK163280.1 | - | 13 | 1323 | |||
| AK166236.1 | - | 13 | 1435 | |||
| AK166834.1 | - | 3 | 3340 | |||
| AK170160.1 | - | 13 | 1447 | |||
| AK210571.1 | - | 3 | 237 | |||
| AK217071.1 | - | 3 | 248 | |||
| BC008103.1 | - | 13 | 1413 | 3966815 | ||
| BC108354.1 | - | 13 | 1397 | 5035560 | ||

Msto1
Genomic Map