siRNA Details
| Gene Symbol: Adamtsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301705.1 | NP_001288634.1 | 5 | 738 | ||
| NM_144899.3 | NP_659148.2 | 5 | 620 | |||
| GenBank mRNA | AK155481.1 | - | 5 | 668 | ||
| AY158701.1 | - | 5 | 561 | |||
| BC117925.1 | - | 4 | 533 | 40097608 | ||

Adamtsl4
Genomic Map