siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178080.4 | NP_835181.2 | 1 | 644 | ||
GenBank mRNA | AK028810.1 | - | 1 | 612 | ||
AK042081.1 | - | 1 | 620 | |||
AK122286.1 | - | 1 | 639 | |||
BC025653.1 | - | 1 | 570 | |||
BC050783.1 | - | 1 | 598 | |||
BC057350.1 | - | 1 | 561 | |||
BC141172.1 | - | 1 | 572 |
