siRNA Details
| Gene Symbol: Vangl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177545.5 | NP_808213.2 | 4 | 963 | ||
| GenBank mRNA | AF481860.1 | - | 4 | 879 | ||
| AK135230.1 | - | 4 | 924 | |||
| AK167743.1 | - | 4 | 845 | |||
| BC138519.1 | - | 3 | 705 | 8861539 | ||
| BC138520.1 | - | 3 | 705 | 8861540 | ||

Vangl1
Genomic Map