siRNA Details
Gene Symbol: St7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253702.1 | NP_001240631.1 | 3 | 977 | ||
NM_001253703.1 | NP_001240632.1 | 3 | 977 | |||
NM_153091.3 | NP_694731.1 | 3 | 977 | |||
GenBank mRNA | AB084518.1 | - | 3 | 391 | ||
AK033865.1 | - | 3 | 409 | |||
AK144075.1 | - | 3 | 417 | |||
AK148099.1 | - | 3 | 679 | |||
AK172102.1 | - | 3 | 978 | |||
AK172218.1 | - | 3 | 978 | |||
BC024473.1 | - | 3 | 368 | |||
BC058781.1 | - | 3 | 402 | 6311899 | ||
BC107394.2 | - | 3 | 402 | 40058131 | ||
BC145950.1 | - | 3 | 469 | 40131253 |
