siRNA Details
| Gene Symbol: AI504432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039146.1 | - | 2 | 338 | ||
| BC127040.1 | - | 2 | 338 | |||
| BC127041.1 | - | 2 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI504432| Gene Symbol: AI504432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039146.1 | - | 2 | 338 | ||
| BC127040.1 | - | 2 | 338 | |||
| BC127041.1 | - | 2 | 338 | |||