siRNA Details
| Gene Symbol: Slc16a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146136.1 | NP_666248.1 | 4 | 444 | ||
| GenBank mRNA | AK045797.1 | - | 4 | 430 | ||
| AK049873.1 | - | 4 | 507 | |||
| AK052779.1 | - | 4 | 443 | |||
| AK085492.1 | - | 4 | 441 | |||
| AK136837.1 | - | 4 | 443 | |||
| BC025441.1 | - | 4 | 432 | 5043709 | ||
| BC026596.1 | - | 4 | 444 | 4975010 | ||

Slc16a4
Genomic Map