siRNA Details
| Gene Symbol: Gpr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032627.1 | - | 2 | 1580 | ||
| AK133702.1 | - | 2 | 1579 | |||
| BC116333.2 | - | 1 | 1236 | 40096913 | ||
| BC116334.2 | - | 1 | 1236 | 40096914 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr61| Gene Symbol: Gpr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032627.1 | - | 2 | 1580 | ||
| AK133702.1 | - | 2 | 1579 | |||
| BC116333.2 | - | 1 | 1236 | 40096913 | ||
| BC116334.2 | - | 1 | 1236 | 40096914 | ||