siRNA Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030016.2 | NP_084292.1 | 2 | 1688 | ||
| GenBank mRNA | AK020899.1 | - | 1 | 105 | ||
| AK039588.1 | - | 2 | 1689 | |||
| AK041636.1 | - | 1 | 154 | |||
| AK163561.1 | - | 1 | 103 | |||
| BC127174.1 | - | 1 | 46 | |||

Trmt13
Genomic Map