siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144902.3 | NP_659151.1 | 5, 6 | 843 | ||
| GenBank mRNA | AK029398.1 | - | 5, 6 | 1215 | ||
| AK049484.1 | - | 5, 6 | 843 | |||
| AK161317.1 | - | 5, 6 | 801 | |||
| BC024110.1 | - | 5, 6 | 839 | 5150304 | ||

Slc35a3
Genomic Map