siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144902.3 | NP_659151.1 | 6, 7 | 979 | ||
| GenBank mRNA | AK029398.1 | - | 6, 7 | 1351 | ||
| AK041195.1 | - | 7, 8 | 1099 | |||
| AK049484.1 | - | 6, 7 | 979 | |||
| AK161317.1 | - | 6, 7 | 937 | |||
| BC024110.1 | - | 6, 7 | 975 | 5150304 | ||

Slc35a3
Genomic Map