siRNA Details
| Gene Symbol: Cenpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173762.4 | NP_776123.3 | 39 | 6244 | ||
| GenBank mRNA | AK049676.1 | - | 6 | 1046 | ||
| AY493378.1 | - | 39 | 6157 | |||
| BC049989.1 | - | 5 | 809 | |||
| BC059032.1 | - | 7 | 1206 | |||

Cenpe
Genomic Map