siRNA Details
| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145546.1 | NP_663521.1 | 6 | 795 | ||
| GenBank mRNA | AK131856.1 | - | 6 | 818 | ||
| AK133097.1 | - | 6 | 817 | |||
| AK133825.1 | - | 6 | 817 | |||
| AK143935.1 | - | 6 | 817 | |||
| AK145403.1 | - | 6 | 817 | |||
| AK146223.1 | - | 6 | 818 | |||
| AK146666.1 | - | 6 | 817 | |||
| AK146910.1 | - | 6 | 817 | |||
| AK151577.1 | - | 6 | 817 | |||
| BC016637.1 | - | 6 | 795 | 2650779 | ||

Gtf2b
Genomic Map