siRNA Details
| Gene Symbol: Gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145546.1 | NP_663521.1 | 2 | 120 | ||
| GenBank mRNA | AK131856.1 | - | 2 | 142 | ||
| AK133097.1 | - | 2 | 142 | |||
| AK133825.1 | - | 2 | 142 | |||
| AK143935.1 | - | 2 | 142 | |||
| AK145403.1 | - | 2 | 142 | |||
| AK146223.1 | - | 2 | 143 | |||
| AK146666.1 | - | 2 | 142 | |||
| AK146910.1 | - | 2 | 142 | |||
| AK151577.1 | - | 2 | 142 | |||
| BC016637.1 | - | 2 | 120 | 2650779 | ||

Gtf2b
Genomic Map