siRNA Details
| Gene Symbol: Znhit6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081094.1 | NP_001074563.1 | 1 | 668 | ||
| GenBank mRNA | AK148729.1 | - | 1 | 645 | ||
| AK167835.1 | - | 2 | 90 | |||
| AK168844.1 | - | 2 | 84 | |||
| BC147498.1 | - | 1 | 554 | 9056360 | ||
| BC150966.1 | - | 1 | 554 | 9087878 | ||
| BC150967.1 | - | 1 | 554 | 9087879 | ||
| BC151198.1 | - | 1 | 554 | 9088086 | ||

Znhit6
Genomic Map