siRNA Details
| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172689.3 | NP_766277.3 | 9 | 1405 | ||
| GenBank mRNA | AK052595.1 | - | 5 | 659 | ||
| AK128929.1 | - | 9 | 1340 | |||
| AK155930.1 | - | 9 | 1341 | |||
| AY553221.1 | - | 9 | 1281 | |||
| BC027369.1 | - | 9 | 1384 | |||
| BC106095.1 | - | 9 | 1325 | |||
| BC127163.1 | - | 9 | 1271 | |||
| BC152540.1 | - | 9 | 1273 | |||

Ddx58
Genomic Map